Learn More
Enzo Life Sciences Cathepsin K drug discovery kit (1x96wells)
SDP
Catalog No. 502010280
Encompass_Preferred
The Cathepsin K Drug Discovery Kit is a complete assay system designed to screen inhibitors of cathepsin K. Using a convenient microplate format, cathepsin K activity is monitored fluorimetrically using the substrate provided in the kit. Applications: Fluorescent detection, HTS Activity assay.
Catalog No. 50-201-0280 Supplier Enzo Life Sciences Supplier No. BMLAK4300001
The Cathepsin K Drug Discovery Kit is a complete assay system designed to screen inhibitors of cathepsin K. Using a convenient microplate format, cathepsin K activity is monitored fluorimetrically using the substrate provided in the kit. Applications: Fluorescent detection, HTS Activity assay.