Learn More
Medchemexpress LLC Cathepsin K inhibitor 6 | 325775-42-6 | 98.2% | 251.30 | 25 MG
SDP
Catalog No. 5000363675
Encompass_Preferred
Cathepsin K inhibitor 6 is an inhibitor of cathepsin K (Cat K) with an IC50 of 17 nM. It also exhibits inhibitory effects on cathepsin L (IC50: 0.05 μM) and cathepsin B (IC50: 0.3 μM).
  • Solid appearance
  • White to off-white color
  • Inhibits cathepsin K with an IC50 of 17 nM
  • Inhibits cathepsin L with an IC50 of 0.05 μM
  • Inhibits cathepsin B with an IC50 of 0.3 μM
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month
Catalog No. 50-003-63675 Supplier Medchemexpress LLC Supplier No. 25MGHY147774
Cathepsin K inhibitor 6 is an inhibitor of cathepsin K (Cat K) with an IC50 of 17 nM. It also exhibits inhibitory effects on cathepsin L (IC50: 0.05 μM) and cathepsin B (IC50: 0.3 μM).
  • Solid appearance
  • White to off-white color
  • Inhibits cathepsin K with an IC50 of 17 nM
  • Inhibits cathepsin L with an IC50 of 0.05 μM
  • Inhibits cathepsin B with an IC50 of 0.3 μM
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month