Learn More
Medchemexpress LLC Cathepsin K Inhibitor 6 | 325775-42-6 | 98.2% | 251.30 | 5 MG
SDP
Catalog No. 5000362224
Encompass_Preferred
Cathepsin K inhibitor 6 is a potent inhibitor of cathepsin K (IC50 17 nM), a key enzyme. It also demonstrates inhibitory effects on other cathepsin isoforms, specifically cathepsin L (IC50 0.05 μM) and cathepsin B (IC50 0.3 μM), making it a valuable tool for research in metabolic enzyme and protease pathways.
  • Inhibits cathepsin K with an IC50 of 17 nM
  • Inhibits cathepsin L with an IC50 of 0.05 μM
  • Inhibits cathepsin B with an IC50 of 0.3 μM
  • Purity of 98.18%
  • Appears as a white to off-white solid
  • Suitable for in vitro and in vivo studies
Catalog No. 50-003-62224 Supplier Medchemexpress LLC Supplier No. HY1477745MG
Cathepsin K inhibitor 6 is a potent inhibitor of cathepsin K (IC50 17 nM), a key enzyme. It also demonstrates inhibitory effects on other cathepsin isoforms, specifically cathepsin L (IC50 0.05 μM) and cathepsin B (IC50 0.3 μM), making it a valuable tool for research in metabolic enzyme and protease pathways.
  • Inhibits cathepsin K with an IC50 of 17 nM
  • Inhibits cathepsin L with an IC50 of 0.05 μM
  • Inhibits cathepsin B with an IC50 of 0.3 μM
  • Purity of 98.18%
  • Appears as a white to off-white solid
  • Suitable for in vitro and in vivo studies