Learn More
Medchemexpress LLC Cathepsin K inhibitor 6 | 325775-42-6 | 98.2% | 251.30 | 50 MG
SDP
Catalog No. 5000361595
Encompass_Preferred
Cathepsin K inhibitor 6 (compound 19) is an inhibitor of cathepsin K (Cat K) with an IC50 of 17 nM. It also inhibits other isoforms, with IC50s of 0.05 μM (Cat L) and 0.3 μM (Cat B).
  • Inhibitor of cathepsin K (Cat K)
  • IC50: 17 nM (Cat K)
  • IC50: 0.05 μM (Cat L)
  • IC50: 0.3 μM (Cat B)
Catalog No. 50-003-61595 Supplier Medchemexpress LLC Supplier No. HY14777450MG
Add to Cart
Add to Cart
Cathepsin K inhibitor 6 (compound 19) is an inhibitor of cathepsin K (Cat K) with an IC50 of 17 nM. It also inhibits other isoforms, with IC50s of 0.05 μM (Cat L) and 0.3 μM (Cat B).
  • Inhibitor of cathepsin K (Cat K)
  • IC50: 17 nM (Cat K)
  • IC50: 0.05 μM (Cat L)
  • IC50: 0.3 μM (Cat B)