Learn More
Enzo Life Sciences Cathepsin K (human), (recombinant) (10 µg)
SDP
Catalog No. NC9062521
Encompass_Preferred
Highly active cysteine protease. Alternative name: Cathepsin O. Cathepsin O2. MW: ~26kDa. Concentration: 0.023mg/ml. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM sodium acetate, pH 5.5, containing 50mM sodium chloride, 0.5mM EDTA and 5mM DTT. Source: Produced in insect cells. Purified as full-length proenzyme (see Prod. No. BML-SE367), then auto-activated at low pH. Produced in a baculovirus expression system. Use/Stability: Stable for at least 6 months after receipt when stored at -80°C. UniProt: P43235. Handling: Keep on dry ice. Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C. Long Term Storage: -80°C
Catalog No. NC9062521 Supplier Enzo Life Sciences Supplier No. BMLSE5530010
Highly active cysteine protease. Alternative name: Cathepsin O. Cathepsin O2. MW: ~26kDa. Concentration: 0.023mg/ml. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM sodium acetate, pH 5.5, containing 50mM sodium chloride, 0.5mM EDTA and 5mM DTT. Source: Produced in insect cells. Purified as full-length proenzyme (see Prod. No. BML-SE367), then auto-activated at low pH. Produced in a baculovirus expression system. Use/Stability: Stable for at least 6 months after receipt when stored at -80°C. UniProt: P43235. Handling: Keep on dry ice. Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C. Long Term Storage: -80°C