Learn More
Athens Research And Technology Inc Cathepsin L, Native Human Liver, 25 µg
Catalog No. 50489704
Encompass_Preferred
Cathepsin L is a native protein purified from human liver, supplied frozen in a 25 µg pack. Molecular weight: 25 - 29 kDa. Cathepsin L, Human Liver: 25 UG; Frozen in 20 mM Malonate, pH 5.5, 1 mM EDTA, 400 mM NaCl. MW 25,000-29,000; Extinction Coefficient=1.28; Common research applications include protease activity, enzyme characterization and biochemical studies.
Catalog No. 50-489-704 Supplier Athens Research And Technology Inc Supplier No. 161203011225UG
Cathepsin L is a native protein purified from human liver, supplied frozen in a 25 µg pack. Molecular weight: 25 - 29 kDa. Cathepsin L, Human Liver: 25 UG; Frozen in 20 mM Malonate, pH 5.5, 1 mM EDTA, 400 mM NaCl. MW 25,000-29,000; Extinction Coefficient=1.28; Common research applications include protease activity, enzyme characterization and biochemical studies.