Learn More
Cayman Chemical CathepsIn L InhIbItr 25mg
SDP
Catalog No. 502986939
Encompass_Preferred
A potent cathepsin L inhibitor (IC50 = 0.85 nM); selective for cathepsin L over calpain II and cathepsin B (IC50s = 184, and 85.1 nM, respectively); trypanocidal against T. brucei (ED50 = 45 nM); completely suppresses osteoclastic pit formation on femur slices isolated from bovine cortical bone at a concentration of 1 μg/ml; inhibits bone loss in a mouse model of osteoporosis in a dose-dependent manner
Catalog No. 50-298-6939 Supplier Cayman Chemical Supplier No. 2324925 MG
A potent cathepsin L inhibitor (IC50 = 0.85 nM); selective for cathepsin L over calpain II and cathepsin B (IC50s = 184, and 85.1 nM, respectively); trypanocidal against T. brucei (ED50 = 45 nM); completely suppresses osteoclastic pit formation on femur slices isolated from bovine cortical bone at a concentration of 1 μg/ml; inhibits bone loss in a mouse model of osteoporosis in a dose-dependent manner