Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cathepsin L/V/K/H Antibody (YA2547) | 50 UL
SDP

Catalog No. 5000443950
Encompass_Preferred

Cathepsin L/V/K/H Antibody (YA2547) is a rabbit-derived and non-conjugated IgG monoclonal antibody, targeting cathepsin L/V/K/H. This antibody is supplied in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol, and is intended for research use only.

  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets cathepsin L/V/K/H
  • Species reactivity includes human, mouse, and rat
  • Immunogen is a synthesized peptide derived from human cathepsin L/V/K/H aa110-135
  • Suitable for applications like western blot (WB), immunohistochemistry-paraffin (IHC-P), immunocytochemistry/immunofluorescence (ICC/IF), and flow cytometry (FC)
  • Predicted and observed band size: 38 kDa
  • Purity is achieved via affinity chromatography
  • Isotype: IgG
  • Clonality: recombinant, monoclonal
  • Endogenous sensitivity
  • Store at -20°C for 1 year; avoid repeated freeze/thaw cycles

Catalog No. 50-004-43950 Supplier Medchemexpress LLC Supplier No. HYP8280250UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cathepsin L/V/K/H Antibody (YA2547) is a rabbit-derived and non-conjugated IgG monoclonal antibody, targeting cathepsin L/V/K/H. This antibody is supplied in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol, and is intended for research use only.

  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets cathepsin L/V/K/H
  • Species reactivity includes human, mouse, and rat
  • Immunogen is a synthesized peptide derived from human cathepsin L/V/K/H aa110-135
  • Suitable for applications like western blot (WB), immunohistochemistry-paraffin (IHC-P), immunocytochemistry/immunofluorescence (ICC/IF), and flow cytometry (FC)
  • Predicted and observed band size: 38 kDa
  • Purity is achieved via affinity chromatography
  • Isotype: IgG
  • Clonality: recombinant, monoclonal
  • Endogenous sensitivity
  • Store at -20°C for 1 year; avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.