Learn More
Enzo Life Sciences Cathepsin L1 (human), (recombinant) (10µg)
SDP
Catalog No. 502013957
Encompass_Preferred
Alternative name: CTSL1, MEP, CATL, CTSL. Purity: ≥98% (SDS-PAGE). Formulation: Lyophilized from 0.22µm filtered solution in 50mM sodium acetate, pH 5.0, containing 100mM NaCl. Generally 5-8% mannitol or trehalose is added as a protectant before lyophilization. Source: Produced in HEK 293 cells. Long Term Storage: -20°C.
Catalog No. 50-201-3957 Supplier Enzo Life Sciences Supplier No. ENZPRT1690010
Alternative name: CTSL1, MEP, CATL, CTSL. Purity: ≥98% (SDS-PAGE). Formulation: Lyophilized from 0.22µm filtered solution in 50mM sodium acetate, pH 5.0, containing 100mM NaCl. Generally 5-8% mannitol or trehalose is added as a protectant before lyophilization. Source: Produced in HEK 293 cells. Long Term Storage: -20°C.