Learn More
Medchemexpress LLC Cathepsin L1 Human 10ug | 10ug
SDP
Catalog No. 5000187110
Encompass_Preferred
Cathepsin L1 protein is a thiol protease that plays a key role in lysosomal protein degradation affecting cellular functions such as turnover antigen processing and bone remodeling It dissolves cross-linked TG/thyroglobulin releasing the thyroid hormone thyroxine (T4) in the thyroid follicular cavity Cathepsin L1 Protein Human (HEK293 C-His) is the recombinant human-derived Cathepsin L1 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-87110 Supplier Medchemexpress LLC Supplier No. HYP7754A10UG
Cathepsin L1 protein is a thiol protease that plays a key role in lysosomal protein degradation affecting cellular functions such as turnover antigen processing and bone remodeling It dissolves cross-linked TG/thyroglobulin releasing the thyroid hormone thyroxine (T4) in the thyroid follicular cavity Cathepsin L1 Protein Human (HEK293 C-His) is the recombinant human-derived Cathepsin L1 protein expressed by HEK293 with C-6 His labeled tag