Learn More
Sino Biological Cathepsin S / CTSS Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10487T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Cathepsin S / CTSS (rh Cathepsin S / CTSS; Catalog#10487-H08H; NP_004070.3; Met1-Ile331). Cathepsin S / CTSS specific IgG was purified by Human Cathepsin S / CTSS affinity chromatography.
Catalog No. 50-224-8511