Learn More
Sino Biological Cathepsin S / CTSS Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 50769R054100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cathepsin S / CTSS (Catalog#50769-M08H; AAB94925.1; Met1-Ile340).
Catalog No. 50-224-8027