Learn More
Enzo Life Sciences Cathepsin S drug discovery kit (1x96wells)
SDP
Catalog No. 502010281
Encompass_Preferred
A QUANTIZYME Assay System. The Cathepsin S Drug Discovery Kit is a complete assay system designed to screen inhibitors of cathepsin S. Using a convenient microplate format, cathepsin S activity is monitored fluorimetrically using the substrate provided in the kit. Applications: Fluorescent detection, HTS Activity assay.
Catalog No. 50-201-0281 Supplier Enzo Life Sciences Supplier No. BMLAK4310001
A QUANTIZYME Assay System. The Cathepsin S Drug Discovery Kit is a complete assay system designed to screen inhibitors of cathepsin S. Using a convenient microplate format, cathepsin S activity is monitored fluorimetrically using the substrate provided in the kit. Applications: Fluorescent detection, HTS Activity assay.