Learn More
Apexbio Technology LLC Cathepsin S inhibitor 1373215-15-6 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A32845.1
Encompass_Preferred
Cathepsin S inhibitor (CAS 1373215-15-6) is a small-molecule inhibitor targeting the lysosomal cysteine protease Cathepsin S an enzyme critically involved in antigen processing and presentation By reversibly and selectively inhibiting Cathepsin S demonstrating notably less affinity for Cathepsin B this molecule impairs MHC class II-mediated antigen presentation resulting in selective immunosuppressive effects on T cells Consequently Cathepsin S inhibitor holds therapeutic potential for autoimmune and inflammatory conditions such as rheumatoid arthritis psoriasis and multiple sclerosis
Catalog No. 50-000-63150