Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cathepsin Z (CTSZ), mouse | >95.0% | 1 MG
SDP

Catalog No. 5000293420
Encompass_Preferred

Recombinant mouse Cathepsin Z (CTSZ) expressed in HEK293 cells with a C-terminal His tag. The protein exhibits carboxyl monopeptidase and carboxyl dipeptidase activities and is supplied lyophilized for biochemical and enzymatic applications.

  • Species/source: Mouse.
  • Expression system: HEK293 cells.
  • Tag: C-terminal His for affinity purification.
  • Sequence: Mature protein residues A23-V306.
  • Molecular weight: Approximately 38 kDa.
  • Purity: >95.0% by reducing SDS-PAGE.
  • Formulation: Lyophilized from 0.22 μm filtered PBS, pH 7.4.
  • Reconstitution: Do not reconstitute below 100 μg/mL in ddH2O.
  • Storage: Store at -20°C; after reconstitution keep 4°C for 1 week or -20°C for long term.
  • Activity: Reported specific activity 715.46 pmol/min/μg; proenzyme requires activation in acid reducing buffer.

Catalog No. 50-002-93420 Supplier Medchemexpress LLC Supplier No. HYP754481MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse Cathepsin Z (CTSZ) expressed in HEK293 cells with a C-terminal His tag. The protein exhibits carboxyl monopeptidase and carboxyl dipeptidase activities and is supplied lyophilized for biochemical and enzymatic applications.

  • Species/source: Mouse.
  • Expression system: HEK293 cells.
  • Tag: C-terminal His for affinity purification.
  • Sequence: Mature protein residues A23-V306.
  • Molecular weight: Approximately 38 kDa.
  • Purity: >95.0% by reducing SDS-PAGE.
  • Formulation: Lyophilized from 0.22 μm filtered PBS, pH 7.4.
  • Reconstitution: Do not reconstitute below 100 μg/mL in ddH2O.
  • Storage: Store at -20°C; after reconstitution keep 4°C for 1 week or -20°C for long term.
  • Activity: Reported specific activity 715.46 pmol/min/μg; proenzyme requires activation in acid reducing buffer.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.