Learn More
Medchemexpress LLC Cathepsin Z/CTSZ Pro 500ug | 500ug
SDP
Catalog No. 5000187206
Encompass_Preferred
Cathepsin Z/CTSZ Protein exhibits dual carboxy-monopeptidase and carboxy-dipeptidase activities as evidenced by research Additionally it displays functional versatility by generating kinin potentiating peptides Cathepsin Z/CTSZ Protein Human (HEK293 C-His) is the recombinant human-derived Cathepsin Z/CTSZ protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-87206 Supplier Medchemexpress LLC Supplier No. HYP7758A500UG
Cathepsin Z/CTSZ Protein exhibits dual carboxy-monopeptidase and carboxy-dipeptidase activities as evidenced by research Additionally it displays functional versatility by generating kinin potentiating peptides Cathepsin Z/CTSZ Protein Human (HEK293 C-His) is the recombinant human-derived Cathepsin Z/CTSZ protein expressed by HEK293 with C-6 His labeled tag