Learn More
Medchemexpress LLC Cathepsin Z (CTSZ) recombinant protein, mouse | >95.0% | 50 UG

Supplier: Medchemexpress LLC HYP7544850UG
Recombinant mouse Cathepsin Z (CTSZ) is a C-terminal His-tagged protein expressed in HEK293 cells, supplied as a lyophilized powder formulated in PBS, pH 7.4. It displays enzymatic activity suitable for biochemical assays and is provided at high purity for research applications.
- Expressed in HEK293 cells with C-terminal His tag.
- Supplied as lyophilized powder for stable storage and handling.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Specific activity 715.46 pmol/min/μg for enzymatic assays.
- Formulated in PBS, pH 7.4; recommend aliquoting and freezing for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.