Learn More
ACROBiosystems Recombinant Protein;Human Cathepsin B / CTSB Protein;HEK293;50UG;CTB,CTSB,CPSB,Cathepsin B,APPS
Supplier:  ACROBiosystems CTBH522250UG
Encompass_Preferred
Recombinant Protein;Human Cathepsin B / CTSB Protein;50UG;Human Cathepsin B, His Tag (CTB-H5222) is expressed from human 293 cells (HEK293). It contains AA Arg 18 - Ile 339 (Accession # NP_001899).;rh CTSB is fused with a polyhistidine tag at the C-terminus, and has a calculated MW of 36.7 kDa (pro-form) and 29 kDa (mature-form). The predicted N-terminus is Arg18 (pro-form) or Phe74 (mature-form). Due to glycosylation, The reducing (R) protein migrates as 43 kDa (pro-form) band in SDS-PAGE and there may be a 34 kDa (mature-form) band.
Catalog No. 50-165-8979