Learn More
Med Vet International Foley Catheter 5Cc X 22Fr, Each
Catalog No. 502830710
Encompass_Preferred
An innovative manufacturing process provides the maximum amount of silicone coating available. Latex catheter coated internally and externally in 100% silicone to resist encrustation and bacterial colonization. Two-way Foley. Sterile.
Catalog No. 50-283-0710 Supplier Med Vet International Supplier No. FC5X22
An innovative manufacturing process provides the maximum amount of silicone coating available. Latex catheter coated internally and externally in 100% silicone to resist encrustation and bacterial colonization. Two-way Foley. Sterile.