Learn More
Med Vet International Foley Catheter 5Cc X 28Fr, Each
Catalog No. 502830703
Encompass_Preferred
An innovative manufacturing process provides the maximum amount of silicone coating available. Latex catheter coated internally and externally in 100% silicone to resist encrustation and bacterial colonization. Two-way Foley. Sterile.
Catalog No. 50-283-0703 Supplier Med Vet International Supplier No. FC5X28
An innovative manufacturing process provides the maximum amount of silicone coating available. Latex catheter coated internally and externally in 100% silicone to resist encrustation and bacterial colonization. Two-way Foley. Sterile.