Learn More
Sklar Instruments Catheter Instrument Tray, Sklar, 8-7/8 in. x 5 in. x 3 in., Stainless Steel. Sklar Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. EA/1.
SDP
Catalog No. 501437124 Shop All Sklar Instruments Products
Encompass_Preferred
Catheter Instrument Tray, Sklar, 8-7/8 in. x 5 in. x 3 in., Stainless Steel. Sklar Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. Available in a variety of sizes, they feature a seamless design to prevent bacterial growth. Dome or flat lids are also available. The overall size of this catheter tray is 8-7/8 inches x 5 inches x 3 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7124 Supplier Sklar Instruments Supplier No. 101760
Catheter Instrument Tray, Sklar, 8-7/8 in. x 5 in. x 3 in., Stainless Steel. Sklar Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. Available in a variety of sizes, they feature a seamless design to prevent bacterial growth. Dome or flat lids are also available. The overall size of this catheter tray is 8-7/8 inches x 5 inches x 3 inches.