Learn More
Sklar Instruments Catheter Instrument Tray, Sklar, Stainless Steel. 8-7/8 inches x 5 inches x 2 inches. Pack of 6.
SDP
Catalog No. 501437112 Shop All Sklar Instruments Products
Encompass_Preferred
Catheter Instrument Tray, Sklar, Stainless Steel. Sklar's Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. Available in a variety of sizes, they feature a seamless design to prevent bacterial growth. Dome or flat lids are also available. The overall size of this catheter tray is 8-7/8 inches x 5 inches x 2 inches. This sku is for a pack of 6.
This item is not returnable. View return policy
Catalog No. 50-143-7112 Supplier Sklar Instruments Supplier No. 1017386
Catheter Instrument Tray, Sklar, Stainless Steel. Sklar's Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. Available in a variety of sizes, they feature a seamless design to prevent bacterial growth. Dome or flat lids are also available. The overall size of this catheter tray is 8-7/8 inches x 5 inches x 2 inches. This sku is for a pack of 6.