Learn More
Hanna Pharmaceutical Supply Co., Inc. IV CATHETER PERIPHERAL INSYTE AUTOGUARD 22GX1""
Catalog No. NC0304121
Encompass_Preferred
Autoguard™ Needle Shielding DeviceStabilizing wings provide a large surface area for taping down the catheter improving catheter securementWith the push of a button, the needle instantly retracts reducing the risk of accidental needle stick injuriesBD Vialon™, a unique proprietary biomaterial, has been proven to enhance clinical and economic outcomesReduces IV restarts due to reduced complicationsIncreases the ease of penetration 50/BX
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable. View return policy
Catalog No. NC0304121 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. BD381523
 Restricted Item
Add to Cart
Add to Cart
Autoguard™ Needle Shielding DeviceStabilizing wings provide a large surface area for taping down the catheter improving catheter securementWith the push of a button, the needle instantly retracts reducing the risk of accidental needle stick injuriesBD Vialon™, a unique proprietary biomaterial, has been proven to enhance clinical and economic outcomesReduces IV restarts due to reduced complicationsIncreases the ease of penetration 50/BX
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.