Learn More
Med Vet International Jackson Sterile Cat Catheter, 3Fr
Catalog No. 502831514
Encompass_Preferred
This specially designed feline catheter is for aid in treatment of FLUTD. It is a modification of the standard feline catheter but with a shorter length (11cm) and metal stylette. There are convenient holes in the luer mount for suture attachment.
Catalog No. 50-283-1514 Supplier Med Vet International Supplier No. JOR153JA
This specially designed feline catheter is for aid in treatment of FLUTD. It is a modification of the standard feline catheter but with a shorter length (11cm) and metal stylette. There are convenient holes in the luer mount for suture attachment.