Learn More
Med Vet International Canine Urinary Catheter, 4 French, Sterile, J0154Xs
Catalog No. 502829836
Encompass_Preferred
Constructed of a flexible transparent grade of nylon, producing firmness without rigidity. Female luer fitting. Closed end with lateral eye. Sterile.
Catalog No. 50-282-9836 Supplier Med Vet International Supplier No. JOR154XS
Constructed of a flexible transparent grade of nylon, producing firmness without rigidity. Female luer fitting. Closed end with lateral eye. Sterile.