Learn More
Instech Catheter Maintenance Kits: Three Syringes; 1mL Saline (0.9% NaCl), 0.3mL Hep-Glycerol (500 USP units/mL), 1mL syringe (empty), each with PinPort™ Injectors, 50 Kits/150 Syringes each
SDP
Catalog No. NC3920432
Encompass
Prefilled syringes with pharamaceutical-grade solutions you need to streamline your operations. Included is 50 kits containing three syringes each:
  • one with 1 mL of Normal Salline, USP, Sterile Grade for drawing out the lock solution,
  • a second empty syringe to take the blood sample, and
  • a third with 0.3 mL of Heparin Sodium (500 USP units/mL) in Glycerol, USP (50%) lock solution to push back the blood and keep the catheter patent for the next sample.
Unused prefilled syringes may be stored for up to one (1) year from date of manufacture before expiring. The expiration date is listed on the product label.
For laboratory animal research only. Not approved for human use.
Features
  • Preinstalled PinPort™ Injectors; Ready-to-Use with VABs™ and PinPorts™
  • Pharmaceutical-Grade
  • Sterile Filtered
  • Preservative-Free
Catalog No. NC3920432 Supplier Instech Supplier No. USPKS1HGS5000.3NJ1P
Prefilled syringes with pharamaceutical-grade solutions you need to streamline your operations. Included is 50 kits containing three syringes each:
  • one with 1 mL of Normal Salline, USP, Sterile Grade for drawing out the lock solution,
  • a second empty syringe to take the blood sample, and
  • a third with 0.3 mL of Heparin Sodium (500 USP units/mL) in Glycerol, USP (50%) lock solution to push back the blood and keep the catheter patent for the next sample.
Unused prefilled syringes may be stored for up to one (1) year from date of manufacture before expiring. The expiration date is listed on the product label.
For laboratory animal research only. Not approved for human use.
Features
  • Preinstalled PinPort™ Injectors; Ready-to-Use with VABs™ and PinPorts™
  • Pharmaceutical-Grade
  • Sterile Filtered
  • Preservative-Free