Learn More
Instech Catheter for mouse carotid artery, 1 to 3Fr, 8cm, collar @ 1.1cm. Fits 22ga.
SDP
Catalog No. 50890066
Encompass
The Instech C10PU-MCA1301 is a mouse carotid artery catheter featuring a 1Fr polyurethane tip that transitions to 3Fr tubing for a secure fit with 22ga connectors. With an 8cm (3.1in) overall length and a silicone collar positioned at 1.1cm, it provides stable placement during carotid cannulation procedures. The catheter is EtO sterilized, individually pouched, and engineered to deliver low-dead-volume performance for accurate sampling and infusion in small-animal studies.
Features
  • 1Fr to 3Fr polyurethane catheter for mouse carotid artery
  • 8cm (3.1in) overall length
  • 1.1cm collar for vessel anchoring
  • 1Fr segment: 0.18x0.35mm; 3Fr segment: 0.6x1.0mm
  • Fits 22ga connectors and mouse VAB systems
  • Approx. 17µL dead volume
  • EtO sterilized and individually pouched
  • Designed for precise infusion and blood sampling in mice
Catalog No. 50-890-066 Supplier Instech Supplier No. C10PUMCA1301
The Instech C10PU-MCA1301 is a mouse carotid artery catheter featuring a 1Fr polyurethane tip that transitions to 3Fr tubing for a secure fit with 22ga connectors. With an 8cm (3.1in) overall length and a silicone collar positioned at 1.1cm, it provides stable placement during carotid cannulation procedures. The catheter is EtO sterilized, individually pouched, and engineered to deliver low-dead-volume performance for accurate sampling and infusion in small-animal studies.
Features
  • 1Fr to 3Fr polyurethane catheter for mouse carotid artery
  • 8cm (3.1in) overall length
  • 1.1cm collar for vessel anchoring
  • 1Fr segment: 0.18x0.35mm; 3Fr segment: 0.6x1.0mm
  • Fits 22ga connectors and mouse VAB systems
  • Approx. 17µL dead volume
  • EtO sterilized and individually pouched
  • Designed for precise infusion and blood sampling in mice