Learn More
Instech Catheter for mouse femoral vein/artery, 1 to 3Fr, 10.5cm, collar @1.2cm. Fits 22ga.
SDP
Catalog No. 50890067
Encompass
The Instech C10PU-MFV1301 is a mouse femoral vein and artery catheter featuring a 1Fr polyurethane tip that transitions to 3Fr tubing for a secure fit with 22ga connectors. Its 10.5cm (4.1in) length and silicone collar positioned at 1.2cm support stable vessel placement, while the rounded tip improves insertion and long-term patency. Individually pouched and EtO sterilized, it provides low-dead-volume performance for precise infusion and sampling in mouse studies.
Features
  • 1Fr to 3Fr polyurethane catheter for mouse femoral vein or artery
  • 10.5cm (4.1in) overall length
  • 1.2cm silicone collar for vessel anchoring
  • 1Fr segment: 0.18x0.35mm; 3Fr segment: 0.6x1.0mm
  • Fits 22ga mouse VAB connectors
  • Approx. 16µL dead volume
  • Rounded tip for improved patency
  • EtO sterilized and individually pouched
Catalog No. 50-890-067 Supplier Instech Supplier No. C10PUMFV1301
The Instech C10PU-MFV1301 is a mouse femoral vein and artery catheter featuring a 1Fr polyurethane tip that transitions to 3Fr tubing for a secure fit with 22ga connectors. Its 10.5cm (4.1in) length and silicone collar positioned at 1.2cm support stable vessel placement, while the rounded tip improves insertion and long-term patency. Individually pouched and EtO sterilized, it provides low-dead-volume performance for precise infusion and sampling in mouse studies.
Features
  • 1Fr to 3Fr polyurethane catheter for mouse femoral vein or artery
  • 10.5cm (4.1in) overall length
  • 1.2cm silicone collar for vessel anchoring
  • 1Fr segment: 0.18x0.35mm; 3Fr segment: 0.6x1.0mm
  • Fits 22ga mouse VAB connectors
  • Approx. 16µL dead volume
  • Rounded tip for improved patency
  • EtO sterilized and individually pouched