Learn More
Instech Catheter for mouse jugular vein, PU 2Fr 10cm, collar @ 1.3cm, round tip. Fits 25ga.
SDP
Catalog No. 50890082
Encompass
The Instech C20PU-MJV1301 is a mouse jugular vein catheter constructed from 2Fr polyurethane tubing for biocompatibility and a reliable fit with 25ga connection systems. Its 10cm (3.9in) length and silicone collar positioned at 1.3cm support secure placement within the vessel, while the rounded tip is designed to ease insertion and minimize trauma. Individually EtO sterilized, this catheter provides low-dead-volume performance suitable for infusion and sampling applications in mice.
Features
  • 2Fr polyurethane catheter tubing
  • 10cm (3.9in) overall length
  • Silicone collar positioned at 1.3cm
  • Rounded tip for smooth vessel entry
  • 0.43x0.69mm tubing dimensions
  • Fits 25ga mouse VAB connectors
  • Approx. 15µL dead volume
  • Individually manufactured and EtO sterilized
  • Minimum order quantity is 5
Catalog No. 50-890-082 Supplier Instech Supplier No. C20PUMJV1301
The Instech C20PU-MJV1301 is a mouse jugular vein catheter constructed from 2Fr polyurethane tubing for biocompatibility and a reliable fit with 25ga connection systems. Its 10cm (3.9in) length and silicone collar positioned at 1.3cm support secure placement within the vessel, while the rounded tip is designed to ease insertion and minimize trauma. Individually EtO sterilized, this catheter provides low-dead-volume performance suitable for infusion and sampling applications in mice.
Features
  • 2Fr polyurethane catheter tubing
  • 10cm (3.9in) overall length
  • Silicone collar positioned at 1.3cm
  • Rounded tip for smooth vessel entry
  • 0.43x0.69mm tubing dimensions
  • Fits 25ga mouse VAB connectors
  • Approx. 15µL dead volume
  • Individually manufactured and EtO sterilized
  • Minimum order quantity is 5