Learn More
Instech Catheter for rat femoral vein, PU 3Fr 25cm, round tip, collars @ 3.0, 3.5cm. Fits 22ga.
SDP
Catalog No. 50890095
Encompass
The Instech C30PU-RFV1308 is a 3Fr polyurethane catheter designed for rat femoral vein access, providing a reliable 25cm (9.8in) length for infusion and sampling procedures. Its rounded tip facilitates smooth vessel entry, while silicone collars positioned at 3.0cm and 3.5cm support secure vascular anchoring. Manufactured from medical-grade PU and EtO sterilized, this catheter ensures compatibility with 22ga connection systems and maintains low dead volume performance for precise dosing applications.
Features
  • 3Fr polyurethane catheter tubing
  • 25cm (9.8in) overall length
  • Silicone collars at 3.0cm and 3.5cm
  • Rounded tip for improved vessel entry
  • 0.6x1.0mm tubing dimensions
  • Fits 22ga rat VAB and PinPort systems
  • Approx. 75µL dead volume
  • Individually packaged and EtO sterilized
  • Minimum order quantity is 5
Catalog No. 50-890-095 Supplier Instech Supplier No. C30PURFV1308
The Instech C30PU-RFV1308 is a 3Fr polyurethane catheter designed for rat femoral vein access, providing a reliable 25cm (9.8in) length for infusion and sampling procedures. Its rounded tip facilitates smooth vessel entry, while silicone collars positioned at 3.0cm and 3.5cm support secure vascular anchoring. Manufactured from medical-grade PU and EtO sterilized, this catheter ensures compatibility with 22ga connection systems and maintains low dead volume performance for precise dosing applications.
Features
  • 3Fr polyurethane catheter tubing
  • 25cm (9.8in) overall length
  • Silicone collars at 3.0cm and 3.5cm
  • Rounded tip for improved vessel entry
  • 0.6x1.0mm tubing dimensions
  • Fits 22ga rat VAB and PinPort systems
  • Approx. 75µL dead volume
  • Individually packaged and EtO sterilized
  • Minimum order quantity is 5