Learn More
Instech Catheter for rat jugular vein, PU 3Fr 12cm, round tip, collar @ 3.8cm. Fits 22ga.
SDP
Catalog No. 50890111
Encompass
The Instech C30PU-RJV1426 is a 3Fr polyurethane catheter designed for rat jugular vein cannulation, featuring a 12cm (4.7in) length with a rounded distal tip for atraumatic entry. A silicone collar located at 3.8cm supports secure anchoring during implantation, and the catheter is engineered to interface cleanly with 22ga connection systems. Manufactured from medical-grade PU and EtO sterilized, it delivers dependable performance for infusion and sampling applications in rat studies.
Features
  • 3Fr polyurethane catheter tubing
  • 12cm (4.7in) overall length
  • Silicone collar positioned at 3.8cm
  • Rounded tip for smooth vessel entry
  • 0.6x1.0mm tubing dimensions
  • Fits 22ga rat VAB and PinPort systems
  • Individually packaged and EtO sterilized
  • Minimum order quantity is 5
Catalog No. 50-890-111 Supplier Instech Supplier No. C30PURJV1426
The Instech C30PU-RJV1426 is a 3Fr polyurethane catheter designed for rat jugular vein cannulation, featuring a 12cm (4.7in) length with a rounded distal tip for atraumatic entry. A silicone collar located at 3.8cm supports secure anchoring during implantation, and the catheter is engineered to interface cleanly with 22ga connection systems. Manufactured from medical-grade PU and EtO sterilized, it delivers dependable performance for infusion and sampling applications in rat studies.
Features
  • 3Fr polyurethane catheter tubing
  • 12cm (4.7in) overall length
  • Silicone collar positioned at 3.8cm
  • Rounded tip for smooth vessel entry
  • 0.6x1.0mm tubing dimensions
  • Fits 22ga rat VAB and PinPort systems
  • Individually packaged and EtO sterilized
  • Minimum order quantity is 5