Learn More
Instech Catheter for rat jugular vein, PU 3Fr 15cm round tip, collars @ 3.0, 3.5cm from tip, 15mm retention sleeve. Fits 22ga.
SDP
Catalog No. 50890103
Encompass
The Instech C30PU-RJV1309 is a 3Fr polyurethane catheter designed for rat jugular vein cannulation, featuring a 15cm (5.9in) length and a rounded distal tip for smooth vessel entry. Dual silicone collars positioned at 3.0cm and 3.5cm support secure anchoring, while a proximal silicone sleeve provides enhanced connection stability with 22ga systems. Manufactured from medical-grade PU and EtO sterilized, this catheter offers dependable performance for rat IV dosing and sampling applications.
Features
  • 3Fr polyurethane catheter tubing
  • 15cm (5.9in) overall length
  • Silicone collars at 3.0cm and 3.5cm
  • Rounded tip for smooth vessel entry
  • 0.6x1.0mm tubing dimensions
  • Proximal silicone sleeve for secure connections
  • Fits 22ga rat VAB and PinPort systems
  • Approx. 45µL dead volume
  • Individually packaged and EtO sterilized
  • Minimum order quantity is 5
Catalog No. 50-890-103 Supplier Instech Supplier No. C30PURJV1309
The Instech C30PU-RJV1309 is a 3Fr polyurethane catheter designed for rat jugular vein cannulation, featuring a 15cm (5.9in) length and a rounded distal tip for smooth vessel entry. Dual silicone collars positioned at 3.0cm and 3.5cm support secure anchoring, while a proximal silicone sleeve provides enhanced connection stability with 22ga systems. Manufactured from medical-grade PU and EtO sterilized, this catheter offers dependable performance for rat IV dosing and sampling applications.
Features
  • 3Fr polyurethane catheter tubing
  • 15cm (5.9in) overall length
  • Silicone collars at 3.0cm and 3.5cm
  • Rounded tip for smooth vessel entry
  • 0.6x1.0mm tubing dimensions
  • Proximal silicone sleeve for secure connections
  • Fits 22ga rat VAB and PinPort systems
  • Approx. 45µL dead volume
  • Individually packaged and EtO sterilized
  • Minimum order quantity is 5