Learn More
Instech Catheter for rat jugular vein, PU 3Fr 30cm round tip, collars @ 2.5, 3.0cm from tip. Fits 22ga.
SDP
Catalog No. 50890100
Encompass
The Instech C30PU-RJV1303 is a 3Fr polyurethane catheter designed for rat jugular vein cannulation, providing a 30cm (11.8in) length suitable for infusion and sampling workflows. Its rounded tip supports atraumatic vessel entry, and a silicone collar positioned at 2.5cm helps secure placement during surgery. Manufactured from medical-grade PU and EtO sterilized, this catheter is compatible with 22ga connection systems for reliable rat IV studies.
Features
  • 3Fr polyurethane catheter tubing
  • 30cm (11.8in) overall length
  • Silicone collar at 2.5cm
  • Rounded tip for improved vessel entry
  • Fits 22ga rat VAB and PinPort systems
  • Medical-grade PU construction
  • Individually packaged and EtO sterilized
  • Minimum order quantity is 5
Catalog No. 50-890-100 Supplier Instech Supplier No. C30PURJV1303
The Instech C30PU-RJV1303 is a 3Fr polyurethane catheter designed for rat jugular vein cannulation, providing a 30cm (11.8in) length suitable for infusion and sampling workflows. Its rounded tip supports atraumatic vessel entry, and a silicone collar positioned at 2.5cm helps secure placement during surgery. Manufactured from medical-grade PU and EtO sterilized, this catheter is compatible with 22ga connection systems for reliable rat IV studies.
Features
  • 3Fr polyurethane catheter tubing
  • 30cm (11.8in) overall length
  • Silicone collar at 2.5cm
  • Rounded tip for improved vessel entry
  • Fits 22ga rat VAB and PinPort systems
  • Medical-grade PU construction
  • Individually packaged and EtO sterilized
  • Minimum order quantity is 5