Learn More
Med Vet International Suction Catheter 14Fr 5/Pack
Catalog No. 502832340
Encompass_Preferred
These catheters are used when regular suction handles are too large. One end will fit on a standard suction hose. A vent on the catheter allows for on/off suction control. 5 per package.
Catalog No. 50-283-2340 Supplier Med Vet International Supplier No. JOR695C
These catheters are used when regular suction handles are too large. One end will fit on a standard suction hose. A vent on the catheter allows for on/off suction control. 5 per package.