Learn More
Broadley James Corporation CATHODE ASSEMBLY 70 MM DO
Catalog No. NC0047461
Encompass
Cathode Assembly for 25 mm and 19 mm Sensors. This part number includes the cathode assembly and both o-rings required to seal it into the sensor body. The only difference between each of the models listed below is the length of the multi-lead ribbon cable. The ribbon cable connects the assembly to the sensor's 4-pin connector.
Catalog No. NC0047461 Supplier Broadley James Corporation Supplier No. CT25PT050MP
Cathode Assembly for 25 mm and 19 mm Sensors. This part number includes the cathode assembly and both o-rings required to seal it into the sensor body. The only difference between each of the models listed below is the length of the multi-lead ribbon cable. The ribbon cable connects the assembly to the sensor's 4-pin connector.