Learn More
Crescent Chemical Co Inc CATHODE FLUID 10 FOR IEF
SDP
Catalog No. 50248731
Encompass_Preferred
Cathode fluid 10 for IEF Contains 0.22 g L-arginine base, 0.18 g L-lysine base, 6.0 ml ethylene diamine in 50 ml water. Recommended for general use with SERVALYT™ PRECOTES™. Size - 50ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42986.03 CAS
Catalog No. 50-248-731 Supplier Crescent Chemical Co Inc Supplier No. SE42986.03
Cathode fluid 10 for IEF Contains 0.22 g L-arginine base, 0.18 g L-lysine base, 6.0 ml ethylene diamine in 50 ml water. Recommended for general use with SERVALYT™ PRECOTES™. Size - 50ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42986.03 CAS