Learn More
Thermo Scientific Dionex™ Retention Time Standards for Ion Chromatography
Catalog No. NC1734187 Shop All Thermo Scientific Products
Encompass
Standard Components: Lithium 2 mg/L, Sodium 9 mg/L, Ammonium 3 mg/L, Potassium 21 mg/L, Magnesium 9 mg/L, Calcium 17 mg/L | For use with Routine Cation Determinations
Catalog No. NC1734187 Supplier Thermo Scientific Supplier No. 088964
Standard Components: Lithium 2 mg/L, Sodium 9 mg/L, Ammonium 3 mg/L, Potassium 21 mg/L, Magnesium 9 mg/L, Calcium 17 mg/L | For use with Routine Cation Determinations