Learn More
AccuStandard, Inc CATION STANDARD-AMMONIA 500ML
SDP
Supplier:  AccuStandard, Inc ICNH4N1X5
Encompass
Cation Standard - Ammonium as Nitrogen. 500 mL. 100 ?g/mL in 2-5% Nitric Acid, tr Hydrofluoric acid .
Catalog No. NC9337160