Learn More
AccuStandard, Inc CATION STANDARD - AMMONIUM
SDP
Catalog No. NC0830701
Encompass
Cation Standard - Ammonium. 100 mL. 10000 ?g/mL in 2-5% Nitric Acid.
Catalog No. NC0830701 Supplier AccuStandard, Inc Supplier No. ICNH41X1
Cation Standard - Ammonium. 100 mL. 10000 ?g/mL in 2-5% Nitric Acid.