Learn More
Medchemexpress LLC Cationic Copper Phthalocyanine Dye | 33864-99-2 | 100 MG
SDP
Catalog No. 5000410133
Encompass_Preferred
Alcian Blue 8GX is a widely used phthalocyanine dye that demonstrates an affinity for glycoproteins and glycosaminoglycans. Its applications span various biological staining techniques, including the identification of proteins in brain tumors and DNA within cells and tissues.
  • Binds to glycoproteins and glycosaminoglycans.
  • Used for biological staining.
  • Applicable for staining proteins in brain tumors.
  • Used for staining DNA in cells and tissues.
Catalog No. 50-004-10133 Supplier Medchemexpress LLC Supplier No. HYD0001100MG
Alcian Blue 8GX is a widely used phthalocyanine dye that demonstrates an affinity for glycoproteins and glycosaminoglycans. Its applications span various biological staining techniques, including the identification of proteins in brain tumors and DNA within cells and tissues.
  • Binds to glycoproteins and glycosaminoglycans.
  • Used for biological staining.
  • Applicable for staining proteins in brain tumors.
  • Used for staining DNA in cells and tissues.