Learn More
Strem, An Ascensus Company CAS 13965-03-2. 5 x 1vial. trans-Dichlorobis(triphenylphosphine)palladium(II)/potassium phosphate admixture [CatKit single-use vials - 6.32 wt% Pd complex]. MFCD18827649
SDP
Catalog No. 502110846
Encompass
CAS 13965-03-2. 5 x 1vial. trans-Dichlorobis(triphenylphosphine)palladium(II)/potassium phosphate admixture [CatKit single-use vials - 6.32 wt% Pd complex]. MFCD18827649. Molecular Weight 701.89. Molecular Formula PdCl2[P(C6H5)3]2/K3PO4. Color/form off-white pwdr. Strem 46-2038. http//www.strem.com/catalog/v/46-2038/
Catalog No. 50-211-0846 Supplier Strem, An Ascensus Company Supplier No. 4620385VL
CAS 13965-03-2. 5 x 1vial. trans-Dichlorobis(triphenylphosphine)palladium(II)/potassium phosphate admixture [CatKit single-use vials - 6.32 wt% Pd complex]. MFCD18827649. Molecular Weight 701.89. Molecular Formula PdCl2[P(C6H5)3]2/K3PO4. Color/form off-white pwdr. Strem 46-2038. http//www.strem.com/catalog/v/46-2038/