Learn More
Cayman Chemical Catlase asy KIt 96Wel
SDP
Catalog No. 502897025
Encompass_Preferred
Catalase is an ubiquitous antioxidant enzyme that is present in most aerobic cells. It is involved in the detoxification of H2O2, a ROS that is a toxic product of both normal aerobic metabolism and pathogenic ROS production. Cayman’s Catalase Assay utilizes the peroxidatic function of catalase for determination of enzyme activity. The assay can be used to measure catalase activity in plasma, serum, erythrocyte lysates, tissue homogenates, and cell lysates.
Catalog No. 50-289-7025 Supplier Cayman Chemical Supplier No. 70700296 WELL
Add to Cart
Add to Cart
Catalase is an ubiquitous antioxidant enzyme that is present in most aerobic cells. It is involved in the detoxification of H2O2, a ROS that is a toxic product of both normal aerobic metabolism and pathogenic ROS production. Cayman’s Catalase Assay utilizes the peroxidatic function of catalase for determination of enzyme activity. The assay can be used to measure catalase activity in plasma, serum, erythrocyte lysates, tissue homogenates, and cell lysates.