Learn More
CATUG INC CATPURE GAUSSIA LUCIFER 100UG
Catalog No. NC3527168
Encompass
Gaussia Luc mRNA encodes for gaussia luciferase protein,serving as a postitive control for mRNA transfection. Thisproduct is used to both screen and validate delivery systems
Catalog No. NC3527168 Supplier CATUG INC Supplier No. CT084100UG
Gaussia Luc mRNA encodes for gaussia luciferase protein,serving as a postitive control for mRNA transfection. Thisproduct is used to both screen and validate delivery systems