Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ CatScreen™ Rapid Test for Moraxella Cararrhalis, Butyrate Esterase
SDP

Catalog No. 23001817 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Quantity:
25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-001-817 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-817 Supplier Hardy Diagnostics™ Supplier No. Z110
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rapid test for the detection of the enzyme butyrate esterase in bacteria isolated on culture media for the presumptive identification of Branhamella (Moraxella) catarrhalis

  • Performs a butyrate test within 5 minutes
  • Simply moisten disk, inoculate with organism, then wait for the development of a red color
  • For presumptive identification of M. catarrhalis, the CatScreen™ may be used along with the oxidase reaction, gram stain and colony morphology

Specifications

Detectable Analytes Butyrate esterase
Sample Type Moraxella Catarrhalis
Disposable Single-use
No. of Reactions 25
Quantity 25
Analysis Time 5 min.
Form Solid
Operating Temperature Range Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.