Learn More
Medchemexpress LLC HY-N1983 5mg Medchemexpress, Caudatin CAS:38395-02-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN19835MG
Encompass_Preferred
Medchemexpress, HY-N1983 5mg Caudatin CAS:38395-02-7 Caudatin is a steroidal cmpound found in Cynanchum auriculatum, causes cell cycle arrest and induces apoptosis, with anti-cancer and antiangiogenic properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2857