Learn More
Cayman Chemical Caudatn 10mg
SDP
Catalog No. 502946848
Encompass_Preferred
A 5-HT1A receptor mixed antagonist and partial agonist inhibits radioligand binding to 5-HT1A sites in rat hippocampal membranes (Ki 2 nM) decreases 5-carboxamidotryptamine-mediated inhibition of forskolin-induced adenylyl cyclase activity in guinea pig hippocampal membranes antagonizes a1-ARs (pA2s 9.47 9.02 and 9.99 in isolated rat tail artery rabbit aorta and rat aorta respectively) decreases blood pressure in anesthetized rats from 1-300 g/kg inhibits phenylephrine-induced pressor responses in pithed rats potentiates the circadian response to light in dark-adapted and entrained hamsters at 5 mg/kg i.p.
Catalog No. 50-294-6848 Supplier Cayman Chemical Supplier No. 34585 10MG
A 5-HT1A receptor mixed antagonist and partial agonist inhibits radioligand binding to 5-HT1A sites in rat hippocampal membranes (Ki 2 nM) decreases 5-carboxamidotryptamine-mediated inhibition of forskolin-induced adenylyl cyclase activity in guinea pig hippocampal membranes antagonizes a1-ARs (pA2s 9.47 9.02 and 9.99 in isolated rat tail artery rabbit aorta and rat aorta respectively) decreases blood pressure in anesthetized rats from 1-300 g/kg inhibits phenylephrine-induced pressor responses in pithed rats potentiates the circadian response to light in dark-adapted and entrained hamsters at 5 mg/kg i.p.