Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Caustic Individual Test Kit
SDPSCI_ED

Catalog No. S72323 Shop All LaMotte Company Products
Change view
Click to view available options
Range:
1 Drop
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S72323 1 Drop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72323 Supplier LaMotte™ Supplier No. 718101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Safe and accurate tool for testing concentration levels in caustic cleaners

  • Uses drop count method
  • Range and sensitivity is 1 drop = 0.1 or 1% NaOH.
  • Indicator is Phenolphthalein
  • Number of tests is 50 per 10% caustic
  • Ideal for testing sanitation and laundry. A sample is reacted with barium to precipitate any carbonates, then is titrated with a standard acid to phenolphthalein endpoint.

Specifications

For Use With (Application) Laundry and sanitation
Includes 30mL 3.6N hydrochloric acid, 15mL 0.5% phenolphthalein indicator, 30mL 10% barium chloride solution, 0.5mL pipet, plastic, test tube plastic with cap
No. of Tests 50
Range 1 Drop
Type Caustic Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.