Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Caustic Test Kit
SDPSCI_ED

Catalog No. S43594 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
50 tests at 10% (4 reagents)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S43594 50 tests at 10% (4 reagents)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S43594 Supplier LaMotte™ Supplier No. 7516DR02
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • 30g Barium chloride powder
  • 2 x 30mL Hydrochloric acid, 2.5N
  • 60mL Deionized water
  • 15mL Phenolphthalein indicator, 1%
  • 1 Pipet, 1.0mL, plastic
  • 1 Test tube, 5, 10, 15, 20, 25mL, plastic w/ cap
  • 1 Direct reading titrator, 0 to 10 range
  • 1 Spoon, 0.5g, plastic

Caustic test kit for water boiling, cooling and wastewater.

In this caustic test kid, a sample is reacted with barium to precipitate any carbonates, then is titrated with a standard acid to the phenolphthalein endpoint. Range/sensitivity is 0 to10% NaOH.

Specifications

Color Blue
Material Plastic
For Use With (Application) Industrial water boiling and cooling, water and wastewater
Closure Type Screw Cap
Quantity 50 tests at 10% (4 reagents)
Product Type Direct Reading Titrator
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.