Learn More
World Precision Instrument Low Temperature Cautery Kit, 700-1200F
SDP
Catalog No. 50822528
Encompass
For cutting, use the lower voltage with a continuous AC current to product heat rapidly and completely vaporize the tissue at the tip of the probe. For coagulation, use higher voltage in a pulsed manner to produce heat more slowly. With this method, tissue damage is more widespread (a few mm from the probe) and blood coagulates.
Note : For USA order only.
Catalog No. 50-822-528 Supplier World Precision Instrument Supplier No. 501090
For cutting, use the lower voltage with a continuous AC current to product heat rapidly and completely vaporize the tissue at the tip of the probe. For coagulation, use higher voltage in a pulsed manner to produce heat more slowly. With this method, tissue damage is more widespread (a few mm from the probe) and blood coagulates.
Note : For USA order only.