Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ CAUTION Sign

Catalog No. 19129186 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-129-186 Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-129-186 Supplier Brady™ Supplier No. 78063

missing translation for 'validMainframeMsg'

Abrasion-resistant and chemical-resistant

B-836 signs are surface printed and ideal for marking construction sites, temporary parking and asbestos abatement sites

Specifications

Material Polypropylene
Quantity 1 Ea.
For Use With (Application) Safety Awareness Signs
Height (English) 10 in.
Languages English
Dimensions (L x W) 10 x 14 in.
Color Black on Yellow
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.